1-Iodononane
Supplier: Apollo Scientific
Warning
Synonyms: 1-Nonyl iodide
OR5499-25G OR5499-500G
APOSOR5499-25GEA 43.5 EUR
APOSOR5499-25G APOSOR5499-500G
1-Iodononane
1-Iodononane
Formula: C₉H₁₉I
MW: 254.15 g/mol
Boiling Pt: 106…108 °C/8mmHg
Density: 1.288 g/cm³ (25 °C)
Flash Pt: 85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001107
CAS Number: 4282-42-2
EINECS: 224-286-3