Lauric acid 98%

Supplier: Apollo Scientific
Danger

Synonyms: Dodecanoic acid, Dodecylic acid

OR55016-1KG OR55016-25KG OR55016-5KG
APOSOR55016-1KGEA 65.7 EUR
APOSOR55016-1KG APOSOR55016-25KG APOSOR55016-5KG
Lauric acid 98%
Lauric acid
Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
Boiling Pt: 225 °C/100mmHg
Melting Pt: 44…46 °C
Density: 0.883 g/cm³ (25 °C)
Flash Pt: 160–165 °C
Storage Temperature: Ambient
MDL Number: MFCD00002736
CAS Number: 143-07-7
EINECS: 205-582-1
Merck Index: 13,05400

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR