2-Chloro-5-nitrotoluene
Supplier: Apollo Scientific
Synonyms: 1-Chloro-2-methyl-4-nitrobenzene
OR5503-25G OR5503-500G
APOSOR5503-25GEA 0 EUR
APOSOR5503-25G APOSOR5503-500G
2-Chloro-5-nitrotoluene
2-Chloro-5-nitrotoluene
Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 94…96 °C/4mmHg
Melting Pt: 42…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00041430
CAS Number: 13290-74-9