Tetrahydrothiophen-3-one
Supplier: Apollo Scientific
Warning
Synonyms: Dihydrothiophen-3(2H)-one, 4,5-Dihydrothiophen-3(2H)-one, 4,5-Dihydro-3(2H)-thiophenone
OR5506-100G OR5506-25G OR5506-5G
APOSOR5506-100GEA 43.5 EUR
APOSOR5506-100G APOSOR5506-25G APOSOR5506-5G
Tetrahydrothiophen-3-one
Tetrahydrothiophen-3-one
Formula: C₄H₆OS
MW: 102.16 g/mol
Boiling Pt: 175 °C
Density: 1.194 g/cm³ (25 °C)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00005412
CAS Number: 1003-04-9
EINECS: 213-698-9