Citraconic anhydride 99%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Methylfuran-2,5-dione
OR5508-100G OR5508-25G OR5508-250G
APOSOR5508-100GEA 43.5 EUR
APOSOR5508-100G APOSOR5508-25G APOSOR5508-250G
Citraconic anhydride 99%
Citraconic anhydride
Formula: C₅H₄O₃
Boiling Pt: 213…215 °C
Melting Pt: 44475 °C
Density: 1.245 g/cm³ (25 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00005522
CAS Number: 616-02-4
EINECS: 210-459-0
UN: 2922
ADR: 8+6.1,III