(Ethoxycarbonylmethyl)dimethylsulphonium bromide
Supplier: Apollo Scientific
Warning
Synonyms: (2-Ethoxy-2-oxoethyl)(dimethyl)sulphonium bromide
OR5509-10G OR5509-50G
APOSOR5509-10GEA 81.5 EUR
APOSOR5509-10G APOSOR5509-50G
(Ethoxycarbonylmethyl)dimethylsulphonium bromide
(Ethoxycarbonylmethyl)dimethylsulphonium bromide
Formula: CH₃CH₂O₂CCH₂S(CH₃)₂Br
MW: 229.14 g/mol
Melting Pt: 80 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00830557
CAS Number: 5187-82-6
EINECS: 225-968-3