tert-Butyl cyanoacetate
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl 2-cyanoacetate
OR55135-100G OR55135-250G OR55135-25G
APOSOR55135-100GEA 78.3 EUR
APOSOR55135-100G APOSOR55135-250G APOSOR55135-25G
tert-Butyl cyanoacetate
tert-Butyl cyanoacetate
Formula: C₇H₁₁NO₂
MW: 141.17 g/mol
Boiling Pt: 67…68 °C
Density: 0.987 g/cm³ (25 °C)
Flash Pt: 119 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001938
CAS Number: 1116-98-9
EINECS: 214-243-7