4-Chloro-2,6-dimethylpyrimidine 99%
Supplier: Apollo Scientific
Warning
OR55150-25G OR55150-5G OR55150-1G
APOSOR55150-25GEA 88.6 EUR
APOSOR55150-25G APOSOR55150-5G APOSOR55150-1G
4-Chloro-2,6-dimethylpyrimidine 99%
4-Chloro-2,6-dimethylpyrimidine
Formula: C₆H₇ClN₂
MW: 142.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234197
CAS Number: 4472-45-1