4-Hydroxy-3-methoxy-2-nitrobenzaldehyde
Supplier: Apollo Scientific
Danger
OR55157-1G OR55157-5G OR55157-25G
APOSOR55157-1GEA 21.9 EUR
APOSOR55157-1G APOSOR55157-5G APOSOR55157-25G
4-Hydroxy-3-methoxy-2-nitrobenzaldehyde
4-Hydroxy-3-methoxy-2-nitrobenzaldehyde
Formula: C₈H₇NO₅
MW: 197.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017020
CAS Number: 2450-26-2
UN: 2811
ADR: 6.1 III