3-(tert-Butoxycarbonylamino)-1-propanol 95%
Supplier: Apollo Scientific
Warning
Synonyms: 3-(Boc-amino)-1-propanol, N-(3-Hydroxypropyl)carbamic Acid tert-Butyl Ester, Boc-β-Ala-ol, 3-(BOC-amino)-1-propanol, tert-Butyl N-(3-Hydroxypropyl)carbamate
OR55160-100G OR55160-25G
APOSOR55160-100GEA 212 EUR
APOSOR55160-100G APOSOR55160-25G
3-(tert-Butoxycarbonylamino)-1-propanol 95%
3-(tert-Butoxycarbonylamino)-1-propanol
Formula: C₈H₁₇NO₃
Density: 1.025 g/cm³ (25 °C)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00191883
CAS Number: 58885-58-8