5-Bromo-1,3-benzodioxole 95%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromo-1,2-(methylenedioxy)benzene, 5-Bromobenzo[d][1,3]dioxole
OR55178-100G OR55178-500G
APOSOR55178-100GEA 36.4 EUR
APOSOR55178-100G APOSOR55178-500G
5-Bromo-1,3-benzodioxole 95%
5-Bromo-1,3-benzodioxole
Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Boiling Pt: 85…86 °C/1mmHg
Density: 1.669 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00005821
CAS Number: 2635-13-4