4-Pentyn-1-ol 95%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Pentyn-5-ol
OR5518-5G OR5518-25G OR5518-100G
APOSOR5518-5GEA 21.9 EUR
APOSOR5518-5G APOSOR5518-25G APOSOR5518-100G
4-Pentyn-1-ol 95%
4-Pentyn-1-ol
Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 154…155 °C
Density: 0.904 g/cm³ (25 °C)
Flash Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002974
CAS Number: 5390-04-5
EINECS: 226-383-6