4-Hydroxy-2-(methylthio)pyrimidine-5-carboxylic acid
Supplier: Apollo Scientific
Warning
OR55186-1G OR55186-25G OR55186-5G
APOSOR55186-1GEA 139 EUR
APOSOR55186-1G APOSOR55186-25G APOSOR55186-5G
4-Hydroxy-2-(methylthio)pyrimidine-5-carboxylic acid
4-Hydroxy-2-(methylthio)pyrimidine-5-carboxylic acid
Formula: C₆H₆N₂O₃S
MW: 186.190 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09759031
CAS Number: 397308-78-0