tert-Butyl-2,2-dioxooxathiazinane-3-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl 2,2-dioxooxathiazinane-3-carboxylate, tert-Butyl 1,2,3-oxathiazinane-3-carboxylate 2,2-dioxide
OR55220-1G OR55220-250MG OR55220-5G
APOSOR55220-1GEA 364 EUR
APOSOR55220-1G APOSOR55220-250MG APOSOR55220-5G
tert-Butyl-2,2-dioxooxathiazinane-3-carboxylate
tert-Butyl-2,2-dioxooxathiazinane-3-carboxylate
Formula: C₈H₁₅NO₅S
MW: 237.28 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22123271
CAS Number: 521267-18-5