2-Aminoadamantane-2-carboxylic acid
Supplier: Apollo Scientific
Warning
OR55295-250MG OR55295-1G
APOSOR55295-250MGEA 71.2 EUR
APOSOR55295-250MG APOSOR55295-1G
2-Aminoadamantane-2-carboxylic acid
2-Aminoadamantane-2-carboxylic acid
Formula: C₁₁H₁₇NO₂
MW: 195.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09264341
CAS Number: 42381-05-5