1-Bromo-2-butyne 98%
Supplier: Apollo Scientific
Warning
OR5530-25G OR5530-5G OR5530-100G
APOSOR5530-25GEA 31.9 EUR
APOSOR5530-25G APOSOR5530-5G APOSOR5530-100G
1-Bromo-2-butyne 98%
1-Bromo-2-butyne
Formula: C₄H₅Br
Boiling Pt: 123…125 °C
Density: 1.519 g/cm³ (25 °C)
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00190233
CAS Number: 3355-28-0
UN: 1993
ADR: 3,III