Methyl-2-iodo-5-methylbenzoate 97%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 2-Iodo-5-methylbenzoate
OR55475-25G OR55475-5G
APOSOR55475-25GEA 271 EUR
APOSOR55475-25G APOSOR55475-5G
Methyl-2-iodo-5-methylbenzoate 97%
Methyl 2-iodo-5-methylbenzoate
Formula: C₉H₉IO₂
MW: 276.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12198129
CAS Number: 103440-52-4