2'-Methoxy-1'-acetonaphthone 99%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Acetyl-2-methoxynaphthalene, 1-(2-Methoxynaphthalen-1-yl)ethanone
OR55564-1G OR55564-5G
APOSOR55564-1GEA 65.4 EUR
APOSOR55564-1G APOSOR55564-5G
2'-Methoxy-1'-acetonaphthone 99%
2'-Methoxy-1'-acetonaphthone
Formula: C₁₃H₁₂O₂
Melting Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD11553477
CAS Number: 5672-94-6