2,4,6-Tri-tert-butylpyrimidine 99%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2,4,6-Tri-tertbutylpyrimidine
OR55593-1G
OR55593-5G
OR55593-25G
APOSOR55593-1GEA
21.9
EUR
APOSOR55593-1G
APOSOR55593-5G
APOSOR55593-25G
2,4,6-Tri-tert-butylpyrimidine 99%
2,4,6-Tri-tert-butylpyrimidine
Formula:
C₁₆H₂₈N₂ Melting Pt: 77…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094045 CAS Number: 67490-21-5 |