2-Amino-5-bromo-4-methyl-3-nitropyridine 98%
Supplier: Apollo Scientific
Warning
OR5580-10G OR5580-1G OR5580-25G
APOSOR5580-10GEA 187 EUR
APOSOR5580-10G APOSOR5580-1G APOSOR5580-25G
2-Amino-5-bromo-4-methyl-3-nitropyridine 98%
2-Amino-5-bromo-4-methyl-3-nitropyridine
Formula: C₆H₆BrN₃O₂
MW: 232.0400 g/mol
Melting Pt: 160…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00092197
CAS Number: 100367-40-6