2-Acetylpyrazine
Supplier: Apollo Scientific
Warning
Synonyms: Pyrazin-2-ylethan-1-one, Acetylpyrazine
OR5593-100G OR5593-25G OR5593-5G
APOSOR5593-100GEA 75.7 EUR
APOSOR5593-100G APOSOR5593-25G APOSOR5593-5G
2-Acetylpyrazine
2-Acetylpyrazine
Formula: C₆H₆N₂O
Boiling Pt: 78…79 °C/8mmHg
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00006134
CAS Number: 22047-25-2
EINECS: 244-753-5