3-Aminoisoxazole 95%
Supplier: Apollo Scientific
Warning
OR5608-5G OR5608-1G
APOSOR5608-5GEA 43.5 EUR
APOSOR5608-5G APOSOR5608-1G
3-Aminoisoxazole 95%
3-Aminoisoxazole
Formula: C₃H₄N₂O
MW: 84.08 g/mol
Boiling Pt: 226…228 °C
Density: 1.138 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00038814
CAS Number: 1750-42-1