2,4-Thiazolidinedione 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1,3-Thiazolidine-2,4-dione
OR5630-100G OR5630-1KG OR5630-500G
APOSOR5630-100GEA 132 EUR
APOSOR5630-100G APOSOR5630-1KG APOSOR5630-500G
2,4-Thiazolidinedione 98%
2,4-Thiazolidinedione
Formula: C₃H₃NO₂S
MW: 117.13 g/mol
Boiling Pt: 178…179 °C/19mmHg
Melting Pt: 125…127 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005478
CAS Number: 2295-31-0
EINECS: 218-941-2