2,3-Dihydro-6-hydroxymethyl-1H,5H-pyrido[3,2,1-ij]quinolin-5-one 95+%
Supplier: Apollo Scientific
Synonyms: 2,3-Dihydro-6-hydroxymethyl-(1H)-benzo[ij]quinolizin-5-one
OR5632-10G OR5632-1G OR5632-5G
APOSOR5632-10GEA 1340 EUR
APOSOR5632-10G APOSOR5632-1G APOSOR5632-5G
2,3-Dihydro-6-hydroxymethyl-1H,5H-pyrido[3,2,1-ij]quinolin-5-one 95+%
2,3-Dihydro-6-hydroxymethyl-1H,5H-pyrido[3,2,1-ij]quinolin-5-one
Formula: C₁₃H₁₃NO₂
MW: 215.25 g/mol
Melting Pt: 130…132 °C
Storage Temperature: Ambient
MDL Number: MFCD01567811
CAS Number: 386704-11-6