4-Bromoindole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromo-1H-indole
OR5639-25G OR5639-5G OR5639-100G
APOSOR5639-25GEA 128 EUR
APOSOR5639-25G APOSOR5639-5G APOSOR5639-100G
4-Bromoindole 98%
4-Bromoindole
Formula: C₈H₆BrN
MW: 196.05 g/mol
Boiling Pt: 88…90 °C/0.1mmHg
Melting Pt: 17 °C
Density: 1.563 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00671502
CAS Number: 52488-36-5