5-Methyl-2-thiopheneboronic acid 97%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Methylthiophene-2-boronic acid, (5-Methylthiophen-2-yl)boronic acid
OR5665-1G OR5665-5G OR5665-25G
APOSOR5665-1GEA 21.9 EUR
APOSOR5665-1G APOSOR5665-5G APOSOR5665-25G
5-Methyl-2-thiopheneboronic acid 97%
5-Methyl-2-thiopheneboronic acid
Formula: C₅H₇BO₂S
MW: 141.99 g/mol
Melting Pt: 160…165 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD01318166
CAS Number: 162607-20-7