Bis(3-pyridylmethyl)amine
Supplier: Apollo Scientific
Warning
Synonyms: alpha,alpha'-Iminodi(3-picoline), 3,3'-Dipicolylamine
OR5675-1G OR5675-250MG
APOSOR5675-1GEA 85.7 EUR
APOSOR5675-1G APOSOR5675-250MG
Bis(3-pyridylmethyl)amine
Bis(3-pyridylmethyl)amine
Formula: C₁₂H₁₃N₃
Boiling Pt: 168…170 °C/0.3mmHg
Density: 1.12 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00038043
CAS Number: 1656-94-6
EINECS: 216-753-5