3-(1H-Pyrazol-1-yl)benzaldehyde
Supplier: Apollo Scientific
Warning
OR5681-1G OR5681-250MG
APOSOR5681-1GEA 143 EUR
APOSOR5681-1G APOSOR5681-250MG
3-(1H-Pyrazol-1-yl)benzaldehyde
3-(1H-Pyrazol-1-yl)benzaldehyde
Formula: C₁₀H₈N₂O
MW: 172.18 g/mol
Melting Pt: 38…39 °C
Storage Temperature: Ambient
MDL Number: MFCD06740321
CAS Number: 852227-92-0