Ethyl-2-aminothiophene-3-carboxylate 97%
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 2-aminothiophene-3-carboxylate
OR5684-1G OR5684-250MG
APOSOR5684-1GEA 58.2 EUR
APOSOR5684-1G APOSOR5684-250MG
Ethyl-2-aminothiophene-3-carboxylate 97%
Ethyl 2-aminothiophene-3-carboxylate
Formula: C₇H₉NO₂S
MW: 171.22 g/mol
Melting Pt: 47…48 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01566303
CAS Number: 31891-06-2