4-Bromo-N-(3-chloropropyl)benzenesulphonamide 98%
Supplier: Apollo Scientific
OR5703-1G
OR5703-5G
APOSOR5703-1GEA
120
EUR
APOSOR5703-1G
APOSOR5703-5G
4-Bromo-N-(3-chloropropyl)benzenesulphonamide 98%
4-Bromo-N-(3-chloropropyl)benzenesulphonamide
Formula:
C₉H₁₁BrClNO₂S Melting Pt: 88…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD01215119 CAS Number: 98768-71-9 |