2-Chlorobenzyl alcohol 98%
Supplier: Apollo Scientific
Warning
Synonyms: (2-Chlorophenyl)methanol
OR5737-100G OR5737-25G
APOSOR5737-100GEA 24.8 EUR
APOSOR5737-100G APOSOR5737-25G
2-Chlorobenzyl alcohol 98%
2-Chlorobenzyl alcohol
Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 227 °C
Melting Pt: 69…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00004604
CAS Number: 17849-38-6
EINECS: 241-801-7