1-Boc-indole 98%
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl 1-indolecarboxylate
OR5773-100G OR5773-25G
APOSOR5773-100GEA 316 EUR
APOSOR5773-100G APOSOR5773-25G
1-Boc-indole 98%
1-Boc-indole
Formula: C₁₃H₁₅NO₂
MW: 217.27 g/mol
Boiling Pt: 201 °C
Density: 1.07 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD02093939
CAS Number: 75400-67-8