Methyl-3-amino-2-methylbenzoate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 3-amino-2-methylbenzoate, 3-Amino-o-toluic acid Methyl Ester, 3-Amino-2-methylbenzoic acid Methyl Ester
OR5797-100G OR5797-25G
APOSOR5797-100GEA 84.1 EUR
APOSOR5797-100G APOSOR5797-25G
Methyl-3-amino-2-methylbenzoate
Methyl 3-amino-2-methylbenzoate
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 300 °C
Density: 1.146 g/cm³ (25 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00211328
CAS Number: 18583-89-6