3-Methoxycarbonyl-5-methylbenzoic acid 95%
Supplier: Apollo Scientific
|
Warning
|
OR5802-1G
OR5802-5G
APOSOR5802-1GEA
119
EUR
APOSOR5802-1G
APOSOR5802-5G
3-Methoxycarbonyl-5-methylbenzoic acid 95%
3-Methoxycarbonyl-5-methylbenzoic acid
Formula:
C₁₀H₁₀O₄ Melting Pt: 154…158 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07363803 CAS Number: 167299-68-5 |