4,4-Dimethylcyclohexa-1,5-dienylboronic acid pinacol ester
Supplier: Apollo Scientific
Warning
OR5814-1G OR5814-250MG
APOSOR5814-1GEA 757 EUR
APOSOR5814-1G APOSOR5814-250MG
4,4-Dimethylcyclohexa-1,5-dienylboronic acid pinacol ester
4,4-Dimethylcyclohexa-1,5-dienylboronic acid pinacol ester
Formula: C₁₄H₂₃BO₂
Melting Pt: 32…34 °C
Storage Temperature: Freezer
MDL Number: MFCD07363842
CAS Number: 871333-97-0