2-Bromo-1,3,4-thiadiazole 97%
Supplier: Apollo Scientific
Warning
OR5820-1G OR5820-5G OR5820-25G
APOSOR5820-1GEA 58 EUR
APOSOR5820-1G APOSOR5820-5G APOSOR5820-25G
2-Bromo-1,3,4-thiadiazole 97%
2-Bromo-1,3,4-thiadiazole
Formula: C₂HBrN₂S
Melting Pt: 68…73 °C
Storage Temperature: Freezer
MDL Number: MFCD04115375
CAS Number: 61929-24-6