2,5-Dimethylstyrene 97%
Supplier: Apollo Scientific
Warning
OR5826-1G OR5826-5G
APOSOR5826-1GEA 46.6 EUR
APOSOR5826-1G APOSOR5826-5G
2,5-Dimethylstyrene 97%
2,5-Dimethylstyrene
Formula: C₁₀H₁₂
MW: 132.21 g/mol
Boiling Pt: 71…72 °C/10mmHg
Melting Pt: –35 °C
Density: 0.904 g/cm³ (25 °C)
Flash Pt: 64 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008614
CAS Number: 2039-89-6
EINECS: 218-028-9