3-Acetyl-6-bromo-2H-chromen-2-one 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Acetyl-6-bromocoumarin, 3-Acetyl-6-bromo-2H-chromen-2-one
OR5843-10G OR5843-1G OR5843-5G
APOSOR5843-10GEA 87 EUR
APOSOR5843-10G APOSOR5843-1G APOSOR5843-5G
3-Acetyl-6-bromo-2H-chromen-2-one 98%
3-Acetyl-6-bromo-2H-chromen-2-one
Formula: C₁₁H₇BrO₃
MW: 267.09 g/mol
Melting Pt: 227…229 °C
Storage Temperature: Ambient
MDL Number: MFCD00024075
CAS Number: 2199-93-1