2-Aminopyrimidine
Supplier: Apollo Scientific
Warning
Synonyms: Pyrimidin-2-ylamine, Pyrimidin-2-amine
OR5847-100G OR5847-500G
APOSOR5847-100GEA 31.9 EUR
APOSOR5847-100G APOSOR5847-500G
2-Aminopyrimidine
2-Aminopyrimidine
Formula: C₄H₅N₃
MW: 95.1 g/mol
Boiling Pt: 158 °C/186mmHg
Melting Pt: 122…126 °C
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00006089
CAS Number: 109-12-6
EINECS: 203-648-4