(S)-(+)-2-Amino-1-butanol 98%
Supplier: Apollo Scientific
Danger
Synonyms: D(+)-2-Amino-1-butanol, (S)-2-Aminobutan-1-ol
OR5854-100G OR5854-25G OR5854-500G
APOSOR5854-100GEA 142 EUR
APOSOR5854-100G APOSOR5854-25G APOSOR5854-500G
(S)-(+)-2-Amino-1-butanol 98%
(S)-(+)-2-Amino-1-butanol
Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 172…174 °C
Melting Pt: -2
Density: 0.944 g/cm³ (25 °C)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00064418
CAS Number: 5856-62-2
EINECS: 227-475-9
UN: 2735
ADR: 8 III