(R)-(+)-α-Methylbenzylamine
Supplier: Apollo Scientific
Danger
OR5862-100G OR5862-25G
APOSOR5862-100GEA 39.3 EUR
APOSOR5862-100G APOSOR5862-25G
(R)-(+)-α-Methylbenzylamine
(R)-(+)-α-Methylbenzylamine
Formula: C₈H₁₁N
Boiling Pt: 180…186 °C
Melting Pt: –10 °C
Density: 0.952 g/cm³ (25 °C)
Flash Pt: 70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064405
CAS Number: 3886-69-9
EINECS: 223-423-4
UN: 2735
ADR: 8,II