(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol 98%
Supplier: Apollo Scientific
Synonyms: L-(+)-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol, (1S,2S)-(+)-2-Amino-1-(4-nitrophenyl)-1,3-propanediol
OR5869-500G
APOSOR5869-500GEA 102 EUR
APOSOR5869-500G
(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol 98%
(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol
Formula: C₉H₁₂N₂O₄
MW: 212.2026 g/mol
Melting Pt: 163…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00007359
CAS Number: 2964-48-9