(R)-(-)-2-Amino-1-butanol 98%
Supplier: Apollo Scientific
Danger
Synonyms: (R)-2-Aminobutan-1-ol, L(-)-2-Amino-1-butanol
OR5871-100G OR5871-25G OR5871-5G
APOSOR5871-100GEA 198 EUR
APOSOR5871-100G APOSOR5871-25G APOSOR5871-5G
(R)-(-)-2-Amino-1-butanol 98%
(R)-(-)-2-Amino-1-butanol
Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 172…174 °C
Melting Pt: –2 °C
Density: 0.943 g/cm³ (25 °C)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00064419
CAS Number: 5856-63-3
EINECS: 227-476-4
UN: 2735
ADR: 8 III