(S)-(+)-2-Amino-1-propanol
Supplier: Apollo Scientific
Danger
Synonyms: L(+)-Alaninol, L-Alaninol, (S)-2-Aminopropan-1-ol, L(+)-2-Amino-1-propanol
OR5875-100G OR5875-500G OR5875-1KG
APOSOR5875-100GEA 26.1 EUR
APOSOR5875-100G APOSOR5875-500G APOSOR5875-1KG
(S)-(+)-2-Amino-1-propanol
(S)-(+)-2-Amino-1-propanol
Formula: C₃H₉NO
MW: 75.11 g/mol
Boiling Pt: 174…176 °C
Density: 0.965 g/cm³ (25 °C)
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00064412
CAS Number: 2749-11-3
EINECS: 220-388-7
UN: 2735
ADR: 8 II