1,2-Dihydro-4-oxopyrrolo[3,2,1-ij]quinoline-5-carboxaldehyde
Supplier: Apollo Scientific
Warning
OR5883-1G OR5883-250MG
APOSOR5883-1GEA 198 EUR
APOSOR5883-1G APOSOR5883-250MG
1,2-Dihydro-4-oxopyrrolo[3,2,1-ij]quinoline-5-carboxaldehyde
1,2-Dihydro-4-oxopyrrolo[3,2,1-ij]quinoline-5-carboxaldehyde
Formula: C₁₂H₉NO₂
MW: 199.21 g/mol
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD01567793
CAS Number: 386715-47-5