(R)-(-)-2-Amino-3-methyl-1-butanol 98%
Supplier: Apollo Scientific
Warning
Synonyms: D(-)-2-Amino-3-methyl-1-butanol, (R)-(-)-2-Amino-3-methylbutanol, D-Valinol, D(-)-Valinol
OR5885-25G OR5885-100G
APOSOR5885-25GEA 102 EUR
APOSOR5885-25G APOSOR5885-100G
(R)-(-)-2-Amino-3-methyl-1-butanol 98%
(R)-(-)-2-Amino-3-methyl-1-butanol
Formula: C₅H₁₃NO
MW: 103.16 g/mol
Boiling Pt: 189…190 °C
Melting Pt: 35…36 °C
Density: 0.931 g/cm³ (25 °C)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00064297
CAS Number: 4276-09-9