3-Bromo-5-chloro-2-hydroxybenzaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromo-5-chlorosalicylaldehyde
OR5927-25G OR5927-1G OR5927-5G
APOSOR5927-25GEA 87 EUR
APOSOR5927-25G APOSOR5927-1G APOSOR5927-5G
3-Bromo-5-chloro-2-hydroxybenzaldehyde
3-Bromo-5-chloro-2-hydroxybenzaldehyde
Formula: C₇H₄BrClO₂
MW: 235.46 g/mol
Melting Pt: 85…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00051690
CAS Number: 19652-32-5