2-(3-Bromophenyl)-5-methyl-1,3,4-oxadiazole
Supplier: Apollo Scientific
|
Warning
|
OR59322-1G
OR59322-5G
APOSOR59322-1GEA
125
EUR
APOSOR59322-1G
APOSOR59322-5G
2-(3-Bromophenyl)-5-methyl-1,3,4-oxadiazole
2-(3-Bromophenyl)-5-methyl-1,3,4-oxadiazole
Formula:
C₉H₇BrN₂O MW: 239.0700 g/mol Melting Pt: 79…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD08691468 CAS Number: 41491-53-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...