3-Methylbenzo[b]thiophene 96%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Methylbenzothiophene
OR5934-100G
OR5934-25G
APOSOR5934-100GEA
1040
EUR
APOSOR5934-100G
APOSOR5934-25G
3-Methylbenzo[b]thiophene 96%
3-Methylbenzo[b]thiophene
Formula:
C₉H₈S MW: 148.23 g/mol Boiling Pt: 72…74 °C/2mmHg Melting Pt: –40 °C Density: 1.106 g/cm³ (25 °C) Flash Pt: 104 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040243 CAS Number: 1455-18-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...