3-Bromo-2-methoxypyridine 98%
Supplier: Apollo Scientific
Danger
OR59385-1G OR59385-5G OR59385-10G
APOSOR59385-1GEA 21.9 EUR
APOSOR59385-1G APOSOR59385-5G APOSOR59385-10G
3-Bromo-2-methoxypyridine 98%
3-Bromo-2-methoxypyridine
Formula: C₆H₆BrNO
MW: 188.02 g/mol
Boiling Pt: 70…73 °C/3mmHg
Density: 1.58 g/cm³ (25 °C)
Flash Pt: 93.3 °C
Storage Temperature: Ambient
MDL Number: MFCD03095349
CAS Number: 13472-59-8